missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead plate, 6.35mm (0.25 in.) thick, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 12603RF

 View more versions of this product

Catalog No. AA12603RF



Specifications

Specifications

Lead plate
30 x 30cm
7439-92-1
Pb
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
[Pb]
Odorless
5352425
Plate
Gray
≈1.6kg/15x15cm
6.35mm (0.25 in.) thick
(metals basis)
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
lead
207.2
CHEBI:27889
99.9%

Chemical Identifiers

7439-92-1
207.2
WABPQHHGFIMREM-UHFFFAOYSA-N
5352425
lead
Pb
MFCD00134050
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
CHEBI:27889
[Pb]
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : OF7525000

TSCA : TSCA

Documents