missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead plate, 6.35mm (0.25 in.) thick, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 12603VA

 View more versions of this product

Catalog No. AA12603VA



Specifications

Specifications

Lead plate
15 x 15cm
7439-92-1
Pb
blei
[Pb]
Odorless
5352425
Plate
Gray
≈1.6kg/15x15cm
6.35mm (0.25 in.) thick
(metals basis)
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
lead
207.2
CHEBI:27889
99.9%

Chemical Identifiers

7439-92-1
207.2
WABPQHHGFIMREM-UHFFFAOYSA-N
5352425
lead
Pb
MFCD00134050
blei
CHEBI:27889
[Pb]
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c

H350-H360FD-H362-H372

EINECSNumber : 231-100-4

RTECSNumber : OF7525000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents