Learn More
Lead, plasma standard solution, Specpure™, Pb 10,000μg/mL, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 014412AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lead plasma standard solution | |
Pb 10000μg/mL | |
MFCD00134050 | |
Specpure™ |
Pb in 5% HNO3 | |
100 mL | |
UN3264 | |
Liquid |
Chemical Identifiers
MFCD00134050 |
Safety and Handling
GHS H Statement
H360-H314
May damage fertility or the unborn child.
Causes severe skin burns and eye damage.
P201-P202-P234-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335-H350-H360Df-H373
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 231-714-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only