missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13853AD

 View more versions of this product

Catalog No. AA13853AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Lead
Clear liquid
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
lead
5352425
Liquid
Colorless
7439-92-1,7697-37-2,7732-18-5
Pb
Nitric acid: 5.0%; Lead: 0.1%; Water: 94.9%
[Pb]
207.2
CHEBI:27889
23 hPa (17mm Hg) at 20°C
50mL

Chemical Identifiers

7439-92-1
207.2
WABPQHHGFIMREM-UHFFFAOYSA-N
CHEBI:27889
[Pb]
Pb
MFCD00134050
5352425
lead
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

Documents