Please login to your online account to display your discounted pricing

Lead Oxide, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$86.76 - $568.66

Chemical Identifiers

Chemical Name or Material Lead Oxide
CAS 1317-36-8
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula PbO
Formula Weight 223.19
Synonym Litharge
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003364

 
avantor performance
233804
Wide Mouth Amber Glass Bottle 125g Each for $86.76

02003365

 
avantor performance
233805
Poly Bottle 2.5kg Each for $568.66

02003366

 
avantor performance
233801
Poly Bottle 500g Each for $133.07
Description & Specifications

Specifications

Formula Weight 223.19
Chemical Name or Material Lead Oxide
Molecular Formula PbO
Synonym Litharge
Packaging Wide Mouth Amber Glass Bottle
Quantity 125g
Specific Gravity 1L = 9.53kg
Purity Grade BAKER ANALYZED™ Reagent
CAS 1317-36-8