Please login to your online account to display your discounted pricing

Lead Oxide, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  233805

 View more versions of this product

Catalog No. 02003365

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 223.19
Chemical Name or Material Lead Oxide
Synonym Litharge
Molecular Formula PbO
Specific Gravity 1L = 9.53kg
Quantity 2.5kg
Purity Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
CAS 1317-36-8

Specifications

Chemical Name or Material Lead Oxide
CAS 1317-36-8
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula PbO
Formula Weight 223.19
Synonym Litharge