Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead, Oil based standard solution, Specpure™, Pb 1000μg/g

Chemical Identifiers

Chemical Name or Material Lead Oil based standard solution
Name Note Pb 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00134050
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1420818

  SDS
alfa aesar™
1420818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 1 Danger
Chemical Name or Material Lead Oil based standard solution
Physical Form Liquid
Purity Grade Specpure™
Quantity 50g
Health Hazard 3 GHS P Statement
P273-P201-P308+P313
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Name Note Pb 1000μg/g
Health Hazard 2 GHS H Statement
H360-H373
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
MDL Number MFCD00134050
Safety and Handling

Danger

GHS P Statement
P273-P201-P308+P313
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H360-H373
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.