Please login to your online account to display your discounted pricing

Lead Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$131.97 - $478.44

Chemical Identifiers

Chemical Name or Material Lead Nitrate
CAS 10099-74-8
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Pb(NO3)2
Formula Weight 331.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003356

 
avantor performance
232204
Wide Mouth Amber Glass Bottle 125g Each for $131.97

02003357

 
avantor performance
232201
Poly Bottle 500g Each for $478.44
Description & Specifications

Specifications

Physical Form Crystals
Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 331.2
Chemical Name or Material Lead Nitrate
CAS 10099-74-8
Molecular Formula Pb(NO3)2
Quantity 125g
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 4.53kg