Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead naphthenate, 61% min in mineral spirits, Pb 20%

$55.88 - $55.88

Chemical Identifiers

Chemical Name or Material Lead naphthenate
CAS 61790-14-5
MDL Number MFCD00466946
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38593A1

 
alfa aesar™
38593A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3859330

 
alfa aesar™
3859330
250g Each for $55.88
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P210-P260-P261-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Chemical Name or Material Lead naphthenate
TSCA TSCA
Quantity 1kg
Physical Form Liquid
MDL Number MFCD00466946
CAS 61790-14-5
UN Number UN1993
Concentration 61% min. in min.eral spirits, Pb 20%
Health Hazard 1 Danger
Density 1.13g/mL
Health Hazard 2 GHS H Statement
H360-H373-H226-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Color Brown
Flash Point 40° (104°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P261-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.

GHS H Statement
H360-H373-H226-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.