Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead molybdenum oxide, 99%

Chemical Identifiers

Chemical Name or Material Lead molybdenum oxide
CAS 10190-55-3
Molecular Formula MoO4Pb
Linear Formula PbMoO4
Formula Weight 367.13
MDL Number MFCD00016272
UN Number UN3077
Synonym Lead molbydate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1057614

  SDS
alfa aesar™
A1057614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 367.13
Chemical Name or Material Lead molybdenum oxide
TSCA TSCA
Odor Odorless
Assay 99%
Molecular Formula MoO4Pb
Linear Formula PbMoO4
MDL Number MFCD00016272
CAS 10190-55-3
UN Number UN3077
Synonym Lead molbydate
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.