Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead lanthanum zirconium titanium oxide, polymeric precursor

Piezoelectric and electro-optic devices

Chemical Identifiers

Chemical Name or Material Lead lanthanum zirconium titanium oxide
Name Note Polymeric precursor
MDL Number MFCD02684260
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4473403

  SDS
alfa aesar™
4473403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4473406

  SDS
alfa aesar™
4473406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4473414

  SDS
alfa aesar™
4473414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Viscous Liquid
Chemical Name or Material Lead lanthanum zirconium titanium oxide
Name Note Polymeric precursor
Sensitivity Moisture Sensitive
MDL Number MFCD02684260
UN Number UN1993
Color Brown
Quantity 1g
Solubility Information Soluble in hexane, toluene and alcohol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H226-H302-H332-H336
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H226-H302-H332-H336
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.