missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead lanthanum zirconium titanium oxide, polymeric precursor

Piezoelectric and electro-optic devices

Manufacturer:  Alfa Aesar™ 4473406

 View more versions of this product

Catalog No. AA4473406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Lead lanthanum zirconium titanium oxide
MFCD02684260
Soluble in hexane, toluene and alcohol
Moisture Sensitive
5g
Polymeric precursor
UN1993
Viscous Liquid
Brown
Documents