Please login to your online account to display your discounted pricing

Lead(IV) Oxide, For Analysis, EMSURE™, EMD Millipore™


Manufacturer: emd millipore corporation  1074071000

 View more versions of this product

Catalog No. M1074071000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 239.19
DOT Information DOT Class: 5.1 (6.1), Packaging Group: III
Chemical Name or Material Lead(IV) Oxide
EINECS Number 215-174-5
Odor Odorless
Packaging Plastic bottle
Physical Form Solid
Molecular Formula PbO2
Appearance Dark brown solid
CAS 1309-60-0
UN Number UN 1872
Health Hazard 1 H360Df: May damage the unborn child. Suspected of damaging fertility.
H272: May intensify fire; oxidiser.
H302 + H332: Harmful if swallowed or if inhaled
H373: May cause damage to organs through prolonged or repeated exposure.
H410: Very toxic to aquatic life with long lasting effects.

Specifications

Chemical Name or Material Lead(IV) Oxide
CAS 1309-60-0
Molecular Formula PbO2
Formula Weight 239.19
UN Number UN 1872
Accessories

H360Df: May damage the unborn child. Suspected of damaging fertility.
H272: May intensify fire; oxidiser.
H302 + H332: Harmful if swallowed or if inhaled
H373: May cause damage to organs through prolonged or repeated exposure.
H410: Very toxic to aquatic life with long lasting effects.