missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(IV) Oxide, For Analysis, EMSURE™, MilliporeSigma™

Manufacturer:  Emd Millipore Corporation 1074070250

 View more versions of this product

Catalog No. M1074070250



Description

Description

For analysis
Specifications

Specifications

1309-60-0
UN 1872
bioxyde de plomb french, lead brown, lead dioxide, lead iv oxide, lead oxide brown, lead oxide pbo2, lead peroxide, lead peroxide pbo2, lead superoxide
O=[Pb]=O
239.198
239.19
Odorless
250g
O2Pb
Dark brown solid
YADSGOSSYOOKMP-UHFFFAOYSA-N
dioxolead
14793
Solid
Plastic bottle

Chemical Identifiers

1309-60-0
239.198
bioxyde de plomb french, lead brown, lead dioxide, lead iv oxide, lead oxide brown, lead oxide pbo2, lead peroxide, lead peroxide pbo2, lead superoxide
dioxolead
O2Pb
YADSGOSSYOOKMP-UHFFFAOYSA-N
14793
O=[Pb]=O
Safety and Handling

Safety and Handling

H360Df: May damage the unborn child. Suspected of damaging fertility.
H272: May intensify fire; oxidiser.
H302 + H332: Harmful if swallowed or if inhaled
H373: May cause damage to organs through prolonged or repeated exposure.
H410: Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 5.1 (6.1), Packaging Group: III

EINECSNumber : 215-174-5

Documents