missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(IV) Oxide, For Analysis, EMSURE™, MilliporeSigma™

$981.14 - $2,452.71

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.198
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym bioxyde de plomb french
PubChem CID 14793
IUPAC Name dioxolead
SMILES O=[Pb]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1074070250 View Documents Emd Millipore Corporation
1074070250
250g Plastic bottle Each for $981.14
 
M1074071000 View Documents Emd Millipore Corporation
1074071000
1kg Plastic bottle Each for $2,452.71
 
Description

Description

For analysis
Specifications

Chemical Identifiers

1309-60-0
239.198
bioxyde de plomb french
dioxolead
O2Pb
YADSGOSSYOOKMP-UHFFFAOYSA-N
14793
O=[Pb]=O

Specifications

1309-60-0
UN 1872
bioxyde de plomb french
O=[Pb]=O
239.198
239.19
Odorless
250g
O2Pb
Dark brown solid
YADSGOSSYOOKMP-UHFFFAOYSA-N
dioxolead
14793
Solid
Plastic bottle
Lead(IV) Oxide
Safety and Handling

Safety and Handling

H360Df: May damage the unborn child. Suspected of damaging fertility.
H272: May intensify fire; oxidiser.
H302 + H332: Harmful if swallowed or if inhaled
H373: May cause damage to organs through prolonged or repeated exposure.
H410: Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 5.1 (6.1), Packaging Group: III

EINECSNumber : 215-174-5