missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(IV) oxide, Brown to black powder, Puratronic™, 99.995% (Metals basis), Alfa Aesar™

$215.89 - $772.36

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.198
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym bioxyde de plomb french
PubChem CID 14793
IUPAC Name dioxolead
SMILES O=[Pb]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1072909 View Documents Thermo Fisher Scientific
1072909
10g Each for $215.89
 
AA1072918 View Documents Thermo Fisher Scientific
1072918
50g Each for $772.36
 
Specifications

Chemical Identifiers

1309-60-0
239.198
YADSGOSSYOOKMP-UHFFFAOYSA-N
14793
O=[Pb]=O
O2Pb
MFCD00011165
bioxyde de plomb french
dioxolead

Specifications

1309-60-0
O2Pb
bioxyde de plomb french
O=[Pb]=O
239.198
239.19
9.38g/cm3 at 20°C
Brown to Black
Lead(IV) oxide
99.995% (Metals basis)
MFCD00011165
YADSGOSSYOOKMP-UHFFFAOYSA-N
dioxolead
14793
Powder
Odorless
10g
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

H360-H373-H272-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. May intensify fire; oxidizer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 215-174-5