missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Lead(IV) Oxide, ≥97.0%, ACS Reagent, Honeywell™ Fluka™

ACS Reagent,≥97.0%

$207.23 - $532.63

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.198
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead iv oxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
IUPAC Name dioxolead
SMILES O=[Pb]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045460 View Documents Honeywell Fluka™
237140100G
100g Poly Bottle Each for $207.23
 
60045461 View Documents Honeywell Fluka™
237140500G
500g Poly Bottle Each for $532.63
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1309-60-0
239.198
YADSGOSSYOOKMP-UHFFFAOYSA-N
14793
O=[Pb]=O
O2Pb
MFCD00011165
lead dioxide, lead iv oxide, lead peroxide, lead iv oxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
dioxolead

Specifications

1309-60-0
PbO2
UN1872
lead dioxide, lead iv oxide, lead peroxide, lead iv oxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
O=[Pb]=O
239.198
239.2g/mol
ACS Reagent
Black
Lead(Iv) Oxide
O2Pb
MFCD00011165
Powder
YADSGOSSYOOKMP-UHFFFAOYSA-N
dioxolead
14793
≥97.0%
Poly Bottle
100g
Safety and Handling

Safety and Handling

P201-P210-P220-P280-P308 + P313-P370 + P378

H272-H302 + H332-H360Df-H373-H410

DOTInformation : 6.1(5.1)/PGIII

EINECSNumber : 215-174-5

RTECSNumber : OG0700000