Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(IV) fluoride, 99%

Chemical Identifiers

Chemical Name or Material Lead(IV) fluoride
CAS 7783-59-7
Formula Weight 283.18
MDL Number MFCD00016278
UN Number UN2291
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1307004

  SDS
alfa aesar™
1307004
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 283.18
Chemical Name or Material Lead(IV) fluoride
Odor Odorless
Physical Form Crystalline
Assay 99%
Sensitivity Moisture Sensitive
MDL Number MFCD00016278
CAS 7783-59-7
UN Number UN2291
Quantity 2g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.