missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) zirconium oxide, 99.7% (metals basis excluding Hf), Hf 1% max

Manufacturer:  Alfa Aesar™ 1468236

 View more versions of this product

Catalog No. AA1468236



Specifications

Specifications

Lead(II) zirconium oxide
(metals basis excluding Hf), Hf 1% max.
MFCD00016275
lead zirconate, o3zr.pb, dioxido oxo zirconium;lead 2+, $l^ 2-lead 2+ ion oxozirconiumbis olate
[O-][Zr](=O)[O-].[Pb+2]
346.421
346.42
99.7%
−325 Mesh
500g
12060-01-4
O3PbZr
UN3077
RSCSNKLWUCUPDE-UHFFFAOYSA-N
dioxido(oxo)zirconium;lead(2+)
16212542
Powder
7.0g/mL
Odorless

Chemical Identifiers

12060-01-4
346.421
RSCSNKLWUCUPDE-UHFFFAOYSA-N
16212542
[O-][Zr](=O)[O-].[Pb+2]
O3PbZr
MFCD00016275
lead zirconate, o3zr.pb, dioxido oxo zirconium;lead 2+, $l^ 2-lead 2+ ion oxozirconiumbis olate
dioxido(oxo)zirconium;lead(2+)
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

SDS Documents