missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) trifluoroacetate hemihydrate

Manufacturer:  Alfa Aesar™ 1755309

 View more versions of this product

Catalog No. AA1755309



Specifications

Specifications

Lead(II) trifluoroacetate hemihydrate
10g
Odorless
C4F6O4Pb
MFCD00061594
lead trifluoroacetate, lead ii 2,2,2-trifluoroacetate, acmc-1asut, bis trifluoroacetoxy lead ii
C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Pb+2]
433.23
442.23 (433.22 Anhydrous)
147°C to 153°C
4146-73-0
White
Pb(CF3COO)2·0.5H2O
UN2291
XVULNTYAIWEMSW-UHFFFAOYSA-L
lead(2+);2,2,2-trifluoroacetate
71311182
Powder

Chemical Identifiers

4146-73-0
433.23
XVULNTYAIWEMSW-UHFFFAOYSA-L
71311182
C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Pb+2]
C4F6O4Pb
MFCD00061594
lead trifluoroacetate, lead ii 2,2,2-trifluoroacetate, acmc-1asut, bis trifluoroacetoxy lead ii
lead(2+);2,2,2-trifluoroacetate
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

SDS Documents