Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) trifluoroacetate, 98%

Chemical Identifiers

Chemical Name or Material Lead(II) trifluoroacetate
CAS 4146-73-0
Molecular Formula C4F6O4Pb
Linear Formula Pb(CF3COO)2
Formula Weight 433.22g/mol
MDL Number MFCD00061594
UN Number UN2291
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1838109

 
alfa aesar™
A18381-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1838118

 
alfa aesar™
A18381-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 147° to 153°C
Quantity 10g
Odor Odorless
Chemical Name or Material Lead(II) trifluoroacetate
CAS 4146-73-0
Assay 98%
Molecular Formula C4F6O4Pb
Linear Formula Pb(CF3COO)2
Formula Weight 433.22g/mol
MDL Number MFCD00061594
UN Number UN2291
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.