Learn More
Alfa Aesar™ Lead(II) titanium oxide, 99.5%
Manufacturer: Alfa Aesar™ 1413936
Specifications
Lead(II) titanium oxide | |
O3PbTi | |
UN3288 | |
NKZSPGSOXYXWQA-UHFFFAOYSA-N | |
dioxido(oxo)titanium;lead(2+) | |
16211560 | |
Powder | |
7.520g/mL | |
Odorless | |
500g |
12060-00-3 | |
MFCD00011159 | |
lead titanate, lead titanium trioxide, lead ii titanate, lead titanium oxide, lead titanium oxide pbtio3, titanic acid, lead salt, titanox l, titanicacid,leadsalt, o3ti.pb, lead titanate, powder, | |
[O-][Ti](=O)[O-].[Pb+2] | |
303.064 | |
303.09 | |
99.5% | |
−325 Mesh | |
Yellow |
Chemical Identifiers
12060-00-3 | |
303.064 | |
NKZSPGSOXYXWQA-UHFFFAOYSA-N | |
16211560 | |
[O-][Ti](=O)[O-].[Pb+2] |
O3PbTi | |
MFCD00011159 | |
lead titanate, lead titanium trioxide, lead ii titanate, lead titanium oxide, lead titanium oxide pbtio3, titanic acid, lead salt, titanox l, titanicacid,leadsalt, o3ti.pb, lead titanate, powder, | |
dioxido(oxo)titanium;lead(2+) |
Safety and Handling
GHS H Statement
H300-H360-H373-H332
Fatal if swallowed.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
GHS P Statement
P260-P261-P281-P301+P310-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
RTECSNumber : XR1575000
TSCA : TSCA