Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) titanium(IV) 2-ethylhexanoate tetraisopropoxide, 99% (metals basis), 50% w/v in isopropanol

Chemical Identifiers

Chemical Name or Material Lead(II) titanium(IV) 2-ethylhexanoate tetraisopropoxide
Formula Weight 777.4
MDL Number MFCD00171336
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36576AC

  SDS
alfa aesar™
36576AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA36576AE

  SDS
alfa aesar™
36576AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 777.4
Assay Percent Notes (metals basis)
Health Hazard 1 Danger
Quantity 25mL
UN Number UN1993
MDL Number MFCD00171336
Health Hazard 2 GHS H Statement
H225-H360-H373-H302-H319-H336
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration 50% w/v in isopropanol
Chemical Name or Material Lead(II) titanium(IV) 2-ethylhexanoate tetraisopropoxide
Assay 99%
Sensitivity Moisture Sensitive
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Physical Form Liquid
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H360-H373-H302-H319-H336
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes serious eye irritation.
May cause drowsiness or dizziness.