Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) thiosulfate, 97%

Chemical Identifiers

Chemical Name or Material Lead(II) thiosulfate
CAS 13478-50-7
Molecular Formula O3PbS2
Linear Formula PbS2O3
Formula Weight 319.32
MDL Number MFCD00078667
UN Number UN2291
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1938822

  SDS
alfa aesar™
A1938822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1938814

  SDS
alfa aesar™
A1938814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 319.32
CAS 13478-50-7
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Quantity 100g
UN Number UN2291
Odor Odorless
Health Hazard 1 Danger
Chemical Name or Material Lead(II) thiosulfate
Assay 97%
Molecular Formula O3PbS2
TSCA TSCA
Health Hazard 3 GHS P Statement
P273-P201-P501a
Avoid release to the environment.
Obtain special instructions before use.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
MDL Number MFCD00078667
Linear Formula PbS2O3
Safety and Handling

Danger

GHS P Statement
P273-P201-P501a
Avoid release to the environment.
Obtain special instructions before use.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.