Please login to your online account to display your discounted pricing

Lead(II) thiocyanate, 98+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  197030250

 View more versions of this product

Catalog No. AC197030250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Glass bottle
Color White to yellow
Quantity 25g
Chemical Name or Material Lead(II) thiocyanate
CAS 592-87-0
Physical Form White to yellow powder
Assay 98.0% min. (ex Pb)
Assay Percent Range 98+%
Molecular Formula C2N2PbS2
Linear Formula Pb(SCN)2
Formula Weight 323.36g/mol
MDL Number MFCD00011155
Merck Index 15, 5481
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child. Suspected of damaging fertility.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Lead(II) thiocyanate
Assay 98.0% min. (ex Pb)
Assay Percent Range 98+%
Molecular Formula C2N2PbS2
Linear Formula Pb(SCN)2
Formula Weight 323.36g/mol
MDL Number MFCD00011155
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child. Suspected of damaging fertility.
Harmful in contact with skin.

SDS & Safety Documents