Please login to your online account to display your discounted pricing

Lead(II) thiocyanate, 98+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC197030050

 View more versions of this product

Catalog No. AC197030050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0% (ex Pb)
Physical Form Powder
Formula Weight 323.36
Quantity 5g
Color White to yellow
Merck Index 15, 5481
Chemical Name or Material Lead(II) thiocyanate
Packaging Glass bottle
Assay Percent Range 98+%
Linear Formula Pb(SCN)2
CAS 592-87-0
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child. Suspected of damaging fertility.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C2N2PbS2
MDL Number MFCD00011155
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Lead(II) thiocyanate
Assay ≥98.0% (ex Pb)
Assay Percent Range 98+%
Molecular Formula C2N2PbS2
Linear Formula Pb(SCN)2
Formula Weight 323.36
MDL Number MFCD00011155
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child. Suspected of damaging fertility.
Harmful in contact with skin.

SDS & Safety Documents