Learn More
Lead(II) tetrafluoroborate, 50% w/w aq. soln.
Used in Tin-Lead Electroplating | CAS: 13814-96-5 | B2F8Pb | 380.80 g/mol
$398.96 - $882.66
Chemical Identifiers
| CAS | 13814-96-5 |
|---|---|
| Molecular Formula | B2F8Pb |
| Molecular Weight (g/mol) | 380.80 |
| MDL Number | MFCD00016270 |
| InChI Key | UQGXAENNKXXYNF-UHFFFAOYSA-N |
| Synonym | lead ii tetrafluoroborate, lead fluoroborate, lead ii tetrafluoroborate solution, lead boron fluoride, lead tetrafluoroborate, unii-wax1e0vj84, lead ii fluoroborate, acmc-1bpsn, wax1e0vj84, lead 2+ ditetrafluoroborate |
| PubChem CID | 12598180 |
| SMILES | [Pb++].F[B-](F)(F)F.F[B-](F)(F)F |
Description
Used in tin-lead electroplatingLead(II) tetrafluoroborate is used as a catalyst involved in calixarene formation through hydrogen-bonded dimmers and in the preparation of linear polyesters. It is involved in the development and optimization of a bipolar lead-boron tetrafluoride accumulator. It is an electroplating solution for coating metal objects with lead and curing agent for epoxy resins. Further, it is used in printing lines, tin-lead alloy plating, lead soldering and also as a circuit board tin-lead alloy plating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13814-96-5 | |
| 380.80 | |
| UQGXAENNKXXYNF-UHFFFAOYSA-N | |
| 12598180 |
| B2F8Pb | |
| MFCD00016270 | |
| lead ii tetrafluoroborate, lead fluoroborate, lead ii tetrafluoroborate solution, lead boron fluoride, lead tetrafluoroborate, unii-wax1e0vj84, lead ii fluoroborate, acmc-1bpsn, wax1e0vj84, lead 2+ ditetrafluoroborate | |
| [Pb++].F[B-](F)(F)F.F[B-](F)(F)F |
Specifications
| Colorless | |
| Liquid | |
| B2F8Pb | |
| lead ii tetrafluoroborate, lead fluoroborate, lead ii tetrafluoroborate solution, lead boron fluoride, lead tetrafluoroborate, unii-wax1e0vj84, lead ii fluoroborate, acmc-1bpsn, wax1e0vj84, lead 2+ ditetrafluoroborate | |
| UQGXAENNKXXYNF-UHFFFAOYSA-N | |
| lead(2+);ditetrafluoroborate | |
| 12598180 | |
| Odorless | |
| Lead(II) tetrafluoroborate |
| 13814-96-5 | |
| 1 kg | |
| MFCD00016270 | |
| Miscible with water. | |
| [Pb++].F[B-](F)(F)F.F[B-](F)(F)F | |
| 380.80 | |
| 380.81 | |
| 1.62 |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
DOTInformation : DOT Class: 8, Packaging Group: III
EINECSNumber : 237-486-0
RTECSNumber : ED2700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only