Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) tartrate, 99%

Chemical Identifiers

Chemical Name or Material Lead (II) tartrate
CAS 815-84-9
Molecular Formula C4H4O6Pb
Formula Weight 355.27
MDL Number MFCD00043282
UN Number UN2291
Synonym Tartaric acid lead(II) salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1238836

  SDS
alfa aesar™
A1238836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 355.27
TSCA TSCA
Synonym Tartaric acid lead(II) salt
Health Hazard 1 Danger
Chemical Name or Material Lead (II) tartrate
Quantity 500g
Molecular Formula C4H4O6Pb
RTECS Number OG4800000
UN Number UN2291
MDL Number MFCD00043282
CAS 815-84-9
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

Applications
Used as a raw material in organic chemistry for the synthesis of other chiral molecules. The acid has served in the farming and metal industries as a chelating agent for complexing micronutrients.

Solubility
Soluble in dilute nitric acid, alkali, acid ammonium hydrogen oxidation and tartaric acid ammonium solution, insoluble in water and alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.