Learn More
Lead(II) tartrate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals A1238836
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsUsed as a raw material in organic chemistry for the synthesis of other chiral molecules. The acid has served in the farming and metal industries as a chelating agent for complexing micronutrients.
Solubility
Soluble in dilute nitric acid, alkali, acid ammonium hydrogen oxidation and tartaric acid ammonium solution, insoluble in water and alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Lead (II) tartrate | |
2.53 | |
MFCD00043282 | |
UN2291 | |
Soluble in dilute nitric acid,alkali,acid ammonium hydrogen oxidation and tartaric acid ammonium solution,insoluble in water and alcohol. | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.[Pb+2] | |
355.27 | |
355.27 |
815-84-9 | |
C4H4O6Pb | |
500 g | |
unii-x5kmm6t3a6, lead tartrate, x5kmm6t3a6, lead tartrate pbc4h4o6, lead ii tartrate 1:1, lead tartrate van, butanedioic acid, 2,3-dihydroxy-2r,3r-, lead 2+ salt 1:1, butanedioic acid, 2,3-dihydroxy-r-r*,r*-, lead 2+ salt 1:1, +-l-tartaric acid lead ii salt, tartaric acid, lead 2+ salt 1:1 8ci | |
AYPZCTCULRIASE-ZVGUSBNCSA-L | |
(2R,3R)-2,3-dihydroxybutanedioate;lead(2+) | |
101283265 | |
99% |
Chemical Identifiers
815-84-9 | |
355.27 | |
AYPZCTCULRIASE-ZVGUSBNCSA-L | |
101283265 | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.[Pb+2] |
C4H4O6Pb | |
MFCD00043282 | |
unii-x5kmm6t3a6, lead tartrate, x5kmm6t3a6, lead tartrate pbc4h4o6, lead ii tartrate 1:1, lead tartrate van, butanedioic acid, 2,3-dihydroxy-2r,3r-, lead 2+ salt 1:1, butanedioic acid, 2,3-dihydroxy-r-r*,r*-, lead 2+ salt 1:1, +-l-tartaric acid lead ii salt, tartaric acid, lead 2+ salt 1:1 8ci | |
(2R,3R)-2,3-dihydroxybutanedioate;lead(2+) |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
EINECSNumber : 212-426-6
RTECSNumber : OG4800000
TSCA : Yes
Recommended Storage : Ambient temperatures