Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) tantalum oxide, 99.9% (metals basis)

$142.24 - $142.24

Chemical Identifiers

Chemical Name or Material Lead(II) tantalum oxide
CAS 12065-68-8
Formula Weight 665.09
MDL Number MFCD00054256
UN Number UN2291
Synonym Lead(II) tantalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3934814

  SDS
alfa aesar™
3934814
25g Each for $142.24

AA3934822

  SDS
alfa aesar™
3934822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 665.09
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Lead(II) tantalum oxide
TSCA TSCA
Mesh Size −200 Mesh
Physical Form Powder
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00054256
CAS 12065-68-8
UN Number UN2291
Synonym Lead(II) tantalate
Melting Point >350°C
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Density 7.9g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.