Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) sulfide, Puratronic™, 99.995% (metals basis)

Chemical Identifiers

Chemical Name or Material Lead(II) sulfide
CAS 1314-87-0
Purity Grade Puratronic™
Formula Weight 239.25g/mol
MDL Number MFCD00016280
UN Number UN3077
Merck Index 14,5421
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1299704

 
alfa aesar™
12997-04
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 7.5g/mL
Odor Odorless
Refractive Index 3.921
Melting Point 1114°C
Quantity 2g
Chemical Name or Material Lead(II) sulfide
CAS 1314-87-0
Purity Grade Puratronic™
Physical Form Crystalline
Assay 99.995%
Assay Percent Notes (metals basis)
Formula Weight 239.25g/mol
MDL Number MFCD00016280
UN Number UN3077
Merck Index 14,5421
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number OG4550000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.