Please login to your online account to display your discounted pricing

Lead(II) sulfide, 99.995% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Lead(II) sulfide
CAS 1314-87-0
Physical Form Lumps
Assay Percent Range 99.995% (Metal basis)
Molecular Formula PbS
Formula Weight 239.25
MDL Number MFCD00016280
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4546204

  SDS
alfa aesar
4546204
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4546209

  SDS
alfa aesar
4546209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Lumps
Formula Weight 239.25
Quantity 2g
Color Black
DOT Information DOT Class: 9, Packaging Group: III
Chemical Name or Material Lead(II) sulfide
EINECS Number 215-246-6
Odor Odorless
Assay Percent Range 99.995% (Metal basis)
Molecular Formula PbS
Appearance Black lumps
MDL Number MFCD00016280
CAS 1314-87-0
Health Hazard 1 H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
Density 7.5g/cm3 at 20°C
Safety and Handling

H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.