missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) sulfide, 99.995% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4546209

 View more versions of this product

Catalog No. AA4546209



Specifications

Specifications

Lead(II) sulfide
99.995% (Metal basis)
MFCD00016280
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, lead sulfide, natural galena, lead sulphide, natural lead sulfide
S=[Pb]
239.26
239.25
Odorless
10g
1314-87-0
PbS
Black lumps
XCAUINMIESBTBL-UHFFFAOYSA-N
sulfanylidenelead
14819
Lumps
Black

Chemical Identifiers

1314-87-0
239.26
XCAUINMIESBTBL-UHFFFAOYSA-N
14819
S=[Pb]
PbS
MFCD00016280
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, lead sulfide, natural galena, lead sulphide, natural lead sulfide
sulfanylidenelead
Safety and Handling

Safety and Handling

H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 215-246-6

SDS Documents