missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) sulfide, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8828418

 View more versions of this product

Catalog No. AA8828418



Specifications

Specifications

Lead(II) sulfide
99.9% (metals basis)
MFCD00016280
XCAUINMIESBTBL-UHFFFAOYSA-N
sulfanylidenelead
14819
-200 Mesh Powder
1314-87-0
PbS
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, lead sulfide, natural galena, lead sulphide, natural lead sulfide
S=[Pb]
239.26
239.25
50g

Chemical Identifiers

1314-87-0
239.26
XCAUINMIESBTBL-UHFFFAOYSA-N
14819
S=[Pb]
PbS
MFCD00016280
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, lead sulfide, natural galena, lead sulphide, natural lead sulfide
sulfanylidenelead
Safety and Handling

Safety and Handling

EINECSNumber : 215-246-6

SDS Documents