Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) selenate, 99.9%

$102.05 - $102.05

Chemical Identifiers

Chemical Name or Material Lead(II) selenate
CAS 7446-15-3
Formula Weight 350.16
MDL Number MFCD00054013
UN Number UN2630
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3934506

 
alfa aesar™
3934506
5g Each for $102.05

AA3934514

 
alfa aesar™
3934514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 350.16
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Lead(II) selenate
TSCA TSCA
Assay 99.9%
UN Number UN2630
Physical Form Powder
MDL Number MFCD00054013
CAS 7446-15-3
Quantity 5g
Solubility Information Insoluble in water; Soluble in concentrated acids
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H330-H360-H373
Fatal if swallowed.
Fatal if inhaled.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Density 6.37g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENATES
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H360-H373
Fatal if swallowed.
Fatal if inhaled.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.