missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Lead(II) Perchlorate Trihydrate, ACS Reagent, 98%, Honeywell™ Fluka™

ACS Reagent, 98%

$155.95 - $478.48

Chemical Identifiers

CAS 13453-62-8
Molecular Formula Cl2H6O11Pb
Molecular Weight (g/mol) 460.137
MDL Number MFCD00149817
InChI Key KKGGEAQRICYXNM-UHFFFAOYSA-L
Synonym lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent
PubChem CID 25021834
IUPAC Name lead(2+);diperchlorate;trihydrate
SMILES O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Pb+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046724 View Documents Honeywell Fluka™
383066100G
100g Glass Bottle Each for $155.95
 
60046725 View Documents Honeywell Fluka™
383066500G
500g Glass Bottle Each for $478.48
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

13453-62-8
460.137
KKGGEAQRICYXNM-UHFFFAOYSA-L
25021834
O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Pb+2]
Cl2H6O11Pb
MFCD00149817
lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent
lead(2+);diperchlorate;trihydrate

Specifications

13453-62-8
Pb(ClO4)2·3H2O
UN1470
lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent
O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Pb+2]
460.137
460.15g/mol
2.6g/cm3
Glass Bottle
83°C to 84°C
100g
Cl2H6O11Pb
MFCD00149817
Crystalline
KKGGEAQRICYXNM-UHFFFAOYSA-L
lead(2+);diperchlorate;trihydrate
25021834
98%
≤0.005% insolubles
White
3 to 5 (at 25°C)
Lead(II) perchlorate trihydrate
Safety and Handling

Safety and Handling

P201-P210-P220-P280-P308 + P313-P370 + P378

DOTInformation : 6.1(5.1)/PGII

RTECSNumber : 237-125-7