Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) oxide, Puratronic™, 99.9995% (metals basis)

Chemical Identifiers

Chemical Name or Material Lead(II) oxide
CAS 1317-36-8
Purity Grade Puratronic™
Linear Formula PbO
Formula Weight 223.19
MDL Number MFCD00011164
UN Number UN3077
Merck Index 14,5413
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1072814

 
alfa aesar™
1072814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1072822

 
alfa aesar™
1072822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1072836

 
alfa aesar™
1072836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 223.19
Linear Formula PbO
UN Number UN3077
Chemical Name or Material Lead(II) oxide
TSCA TSCA
Odor Odorless
Purity Grade Puratronic™
Physical Form Powder
Assay 99.9995%
Assay Percent Notes (metals basis)
MDL Number MFCD00011164
CAS 1317-36-8
Merck Index 14,5413
Color Yellow
Melting Point 886°C
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 9.53g/mL
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number OG1750000
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.