Please login to your online account to display your discounted pricing

Lead(II) Oxide, Extra Pure, EMD Millipore™


Manufacturer: emd millipore corporation  1056581000

 View more versions of this product

Catalog No. M1056581000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 223.19
DOT Information DOT Class: 6.1, Packaging Group: III
Purity Grade Extra Pure
Odor Odorless
CAS 1317-36-8
Molecular Formula PbO
Packaging Plastic bottle
EINECS Number 215-267-0
Appearance Yellow solid
Chemical Name or Material Lead(II) oxide
Physical Form Solid
UN Number UN 2291
Health Hazard 1 H360FD: May damage fertility. May damage the unborn child.
H302 + H332: Harmful if swallowed or if inhaled
H373: May cause damage to organs through prolonged or repeated exposure.
H410: Very toxic to aquatic life with long lasting effects.

Specifications

Chemical Name or Material Lead(II) oxide
CAS 1317-36-8
Purity Grade Extra Pure
Molecular Formula PbO
Formula Weight 223.19
UN Number UN 2291
Accessories

H360FD: May damage fertility. May damage the unborn child.
H302 + H332: Harmful if swallowed or if inhaled
H373: May cause damage to organs through prolonged or repeated exposure.
H410: Very toxic to aquatic life with long lasting effects.