Learn More
Lead(II) oxide, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 199340025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1470.0°C | |
886.0°C | |
Yellow | |
2.5 kg | |
OPb | |
MFCD00011164 | |
15, 5468 | |
Solubility in water: 70.2mg/l (20°C) | |
O=[Pb] | |
223.20 | |
223.19 | |
Glass bottle |
Lead(II) oxide | |
1317-36-8 | |
Fine Powder | |
99.6% min. (Complexometry) | |
PbO | |
01,536; 02,233 | |
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow | |
YEXPOXQUZXUXJW-UHFFFAOYSA-N | |
plumbanone | |
14827 | |
99.7% |
Chemical Identifiers
1317-36-8 | |
223.20 | |
YEXPOXQUZXUXJW-UHFFFAOYSA-N | |
14827 | |
O=[Pb] |
OPb | |
MFCD00011164 | |
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow | |
plumbanone |
Safety and Handling
GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Causes damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing cancer.
GHS P Statement
Use personal protective equipment as required.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON C
GHS Signal Word: Danger
EINECSNumber : 215-267-
RUO – Research Use Only