missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) oxide, 99.7%, ACROS Organics™

Manufacturer:  Acros Organics 199340010

 View more versions of this product

Catalog No. AC199340010



Specifications

Specifications

Lead(II) oxide
99.7%
PbO
01,536; 02,233
lead ii oxide
YEXPOXQUZXUXJW-UHFFFAOYSA-N
oxolead
250ppm max.
223.19
99.7%
25ppm max.
100ppm max.
20ppm max.
0.1% max.
1470.0°C
886.0°C
1317-36-8
OPb
MFCD00011164
15, 5468
Solubility in water: 70.2mg/l (20°C)
O=[Pb]
223.19
14827
Fine Powder
10ppm max.
200ppm max.
Glass bottle
Solubility in water: 70.2 mg/l (20°C).
0.1% max.
Yellow
1kg

Chemical Identifiers

1317-36-8
223.19
YEXPOXQUZXUXJW-UHFFFAOYSA-N
14827
O=[Pb]
OPb
MFCD00011164
lead ii oxide
oxolead
Safety and Handling

Safety and Handling

GHS P Statement May damage the unborn child. Suspected of damaging fertility. Causes damage to organs through prolonged or repeated exposure. Harmful if inhaled. Harmful if swallowed. Suspected of causing cancer. Very toxic to aquatic life with long lasting effects.

GHS P Statement Use personal protective equipment as required. Wash face, hands and any exposed skin thoroughly after handling. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell.

Danger

SDS Documents