Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) oxalate, Puratronic™, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Lead(II) oxalate
CAS 814-93-7
Purity Grade Puratronic™
Linear Formula PbC2O4
Formula Weight 295.23
MDL Number MFCD00040677
UN Number UN2291
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1299906

 
alfa aesar™
1299906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1299914

 
alfa aesar™
1299914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 295.23
Chemical Name or Material Lead(II) oxalate
TSCA TSCA
Odor Odorless
Purity Grade Puratronic™
Physical Form Powder
Assay 99.999%
Assay Percent Notes (metals basis)
Linear Formula PbC2O4
MDL Number MFCD00040677
CAS 814-93-7
UN Number UN2291
Melting Point 300°C (dec.)
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.