missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) nitrate, White powder, Puratronic™, 99.999% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1072730

 View more versions of this product

Catalog No. AA1072730



Specifications

Specifications

Lead(II) nitrate
99.999% (Metals basis)
MFCD00011153
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead(2+);dinitrate
24924
331.2
Odorless
250g
10099-74-8
N2O6Pb
lead 2+ nitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
331.208
CHEBI:37187
Powder
White

Chemical Identifiers

10099-74-8
331.208
RLJMLMKIBZAXJO-UHFFFAOYSA-N
24924
lead(2+);dinitrate
N2O6Pb
MFCD00011153
lead 2+ nitrate
CHEBI:37187
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
Safety and Handling

Safety and Handling

H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 233-245-9

Documents