missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) nitrate, ACS, 99.0% min, Alfa Aesar™

For Use As An Oxidizer in Dye Industry, in Manufacturing of Mother-of-Pearl, As Mordant in Printing Textile

Manufacturer:  Alfa Aesar 1424336

 View more versions of this product

Catalog No. AA1424336



Specifications

Specifications

Lead(II) nitrate
10099-74-8
N2O6Pb
White crystalline
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead(2+);dinitrate
24924
331.2
ACS
Odorless
White
For ACS analysis
99.0% min.
MFCD00011153
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, lead ii nitrate, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
331.208
CHEBI:37187
Crystals
4.53g/cm3 at 20°C
approx.3.0-4.0 (20% aqueous solution)
500g

Chemical Identifiers

10099-74-8
331.208
RLJMLMKIBZAXJO-UHFFFAOYSA-N
24924
lead(2+);dinitrate
N2O6Pb
MFCD00011153
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, lead ii nitrate, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
CHEBI:37187
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
Safety and Handling

Safety and Handling

H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 233-245-9

Documents