missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lead(II) nitrate, ACS, 99.0% min, Alfa Aesar™
For Use As An Oxidizer in Dye Industry, in Manufacturing of Mother-of-Pearl, As Mordant in Printing Textile
Manufacturer: Alfa Aesar 14243A3
Specifications
Lead(II) nitrate | |
10099-74-8 | |
N2O6Pb | |
White crystalline | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
lead(2+);dinitrate | |
24924 | |
331.2 | |
ACS | |
approx.3.0-4.0 (20% aqueous solution) | |
2kg |
For ACS analysis | |
99.0% min. | |
MFCD00011153 | |
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, lead ii nitrate, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2] | |
331.208 | |
CHEBI:37187 | |
Crystals | |
Odorless | |
White |
Chemical Identifiers
10099-74-8 | |
331.208 | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
24924 | |
lead(2+);dinitrate |
N2O6Pb | |
MFCD00011153 | |
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, lead ii nitrate, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
CHEBI:37187 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2] |
Safety and Handling
H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
DOTInformation : DOT Class: 5.1, Packaging Group: II
EINECSNumber : 233-245-9