missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) nitrate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 211560010

 View more versions of this product

Catalog No. AC211560010


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lead(II) nitrate
10099-74-8
Crystalline Powder
99% min. (Complexometry)
Pb(NO3)2
15,5469
Solubility in water: 343g/L.
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
331.20
CHEBI:37187
≥99%
4.53
470.0°C
White
1 kg
N2O6Pb
MFCD00011153
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead(2+);dinitrate
24924
331.2
Plastic bottle
4.5300g/mL

Chemical Identifiers

10099-74-8
331.20
RLJMLMKIBZAXJO-UHFFFAOYSA-N
24924
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
N2O6Pb
MFCD00011153
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
CHEBI:37187
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
May damage the unborn child.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume

GHS Signal Word: Danger

EINECSNumber : 233-245-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only