Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) metavanadate, 99.9% (metals basis)

As pigment, in manufacturing of vanadium compounds

Chemical Identifiers

Chemical Name or Material Lead(II) metavanadate
CAS 10099-79-3
Formula Weight 405.07
MDL Number MFCD00016271
UN Number UN2291
Merck Index 14,5427
Synonym Lead(V) vanadate; Lead vanadium oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3937818

  SDS
alfa aesar™
3937818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3937830

  SDS
alfa aesar™
3937830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 405.07
Physical Form Powder
TSCA TSCA
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Assay 99.9%
MDL Number MFCD00016271
Merck Index 14,5427
Synonym Lead(V) vanadate; Lead vanadium oxide
Quantity 50g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
Solubility Information Insoluble in water, decomposes in HNO3
Chemical Name or Material Lead(II) metavanadate
Assay Percent Notes (metals basis)
UN Number UN2291
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Odor Odorless
CAS 10099-79-3
Mesh Size −100 Mesh
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.