Please login to your online account to display your discounted pricing

Lead(II,IV) oxide, red, 98%, ACROS Organics™


Quantity 5kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC221110051

 View more versions of this product

Catalog No. AC221110051

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥97.5% (PB3O4)
Formula Weight 685.57
Quantity 5kg
Color Orange
EINECS Number 215-235-6
TSCA TSCA
RTECS Number OG5425000
Chemical Name or Material Lead(II, IV) oxide, red
Magnesium (Mg) 100ppm max.
Name Note 98%
Packaging Glass bottle
Assay Percent Range 98%
Sodium (Na) 100ppm max.
Linear Formula 2PbO·PbO2
MDL Number MFCD00078491
Health Hazard 1 Danger
Molecular Formula O4Pb3
Iron (Fe) 50ppm max.
Manganese (Mn) 10ppm max.
Potassium (K) 100ppm max.
CAS 1314-41-6
Trace Analysis 50ppm max.
Calcium (Ca) 100ppm max.
Synonym Red lead oxide
Health Hazard 2 GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
May damage the unborn child. Suspected of damaging fertility.
Harmful if swallowed.
May intensify fire; oxidizer.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Copper (Cu) 50ppm max.

Specifications

Chemical Name or Material Lead(II, IV) oxide, red
Name Note 98%
CAS 1314-41-6
Assay ≥97.5% (PB3O4)
Assay Percent Range 98%
Molecular Formula O4Pb3
Linear Formula 2PbO·PbO2
Formula Weight 685.57
MDL Number MFCD00078491
Synonym Red lead oxide
Accessories

Danger

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
May damage the unborn child. Suspected of damaging fertility.
Harmful if swallowed.
May intensify fire; oxidizer.

SDS & Safety Documents