missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II,IV) oxide, 97% (metals basis)

Manufacturer:  Alfa Aesar™ 14232A1

 View more versions of this product

Catalog No. AA14232A1



Specifications

Specifications

Lead(II,IV) oxide
(metals basis)
Pb3O4
UN1479
lead tetroxide, orange lead, lead oxide, red, lead ii,iv oxide, po4b3, lead ii,iv oxide, lead oxide, red, lr, lead oxide, red powder, 1-2 mum, lead oxide, red, saj first grade, lead oxide, powder trace metals basis
O1[Pb]O[Pb]12O[Pb]O2
685.596
685.6
97%
Odorless
~500°C (decomposition)
1314-41-6
O4Pb3
MFCD00078491
14,5425
XMFOQHDPRMAJNU-UHFFFAOYSA-N
1,3,5,7-tetraoxa-2$l^{2},4,6$l^{2}-triplumbaspiro[3.3]heptane
16685188
Powder
9.1g/mL
Orange to Red
1kg

Chemical Identifiers

1314-41-6
685.596
XMFOQHDPRMAJNU-UHFFFAOYSA-N
16685188
O1[Pb]O[Pb]12O[Pb]O2
O4Pb3
MFCD00078491
lead tetroxide, orange lead, lead oxide, red, lead ii,iv oxide, po4b3, lead ii,iv oxide, lead oxide, red, lr, lead oxide, red powder, 1-2 mum, lead oxide, red, saj first grade, lead oxide, powder trace metals basis
1,3,5,7-tetraoxa-2$l^{2},4,6$l^{2}-triplumbaspiro[3.3]heptane
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H272-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May intensify fire
oxidizer.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P221-P210-P260-P304+P340-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

RTECSNumber : OG5425000

TSCA : TSCA

Documents