Please login to your online account to display your discounted pricing

Lead(II) iodide, Yellow powder, 99.9985% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Lead(II) iodide
CAS 10101-63-0
Physical Form Powder
Assay Percent Range 99.9985% (Metals basis)
Molecular Formula I2Pb
Formula Weight 461
MDL Number MFCD00011163
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1272414

  SDS
alfa aesar
1272414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1272422

  SDS
alfa aesar
1272422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 461
Physical Form Powder
Quantity 25g
Color Yellow
DOT Information DOT Class: 6.1, Packaging Group: III
EINECS Number 233-256-9
Density 6.16g/cm3 at 20°C
Sensitivity Light sensitive
Odor Odorless
MDL Number MFCD00011163
Health Hazard 2 Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
CAS 10101-63-0
Health Hazard 1 H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
Chemical Name or Material Lead(II) iodide
Molecular Formula I2Pb
Assay Percent Range 99.9985% (Metals basis)
Safety and Handling

H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.