Please login to your online account to display your discounted pricing

Lead(II) iodide, ultra dry, Yellow powder, 99.999% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Lead(II) iodide, ultra dry
CAS 10101-63-0
Purity Grade Ultra dry
Physical Form Powder
Assay Percent Range 99.999% (Metals basis)
Molecular Formula I2Pb
Formula Weight 461
MDL Number MFCD00011163
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4431406

 
alfa aesar
4431406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4431414

 
alfa aesar
4431414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4431422

 
alfa aesar
4431422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Ultra dry
Physical Form Powder
Formula Weight 461
Quantity 5g
Color Yellow
DOT Information DOT Class: 6.1, Packaging Group: III
Chemical Name or Material Lead(II) iodide, ultra dry
EINECS Number 233-256-9
Odor Odorless
Sensitivity Light sensitive
Assay Percent Range 99.999% (Metals basis)
Molecular Formula I2Pb
MDL Number MFCD00011163
CAS 10101-63-0
Health Hazard 1 H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
Health Hazard 2 Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Density 6.16g/cm3 at 20°C
Safety and Handling

H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.